Thermo Scientific Chemicals

Potassium chloride, 0.0375M, Conductivity Standard, Thermo Scientific Chemicals

Catalog number: 042694.K7
4 L, Each
Thermo Scientific Chemicals

Potassium chloride, 0.0375M, Conductivity Standard, Thermo Scientific Chemicals

Catalog number: 042694.K7
4 L, Each
Quantity
Catalog number: 042694.K7
also known as 042694-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7447-40-7
IUPAC Name
potassium chloride
Molecular Formula
ClK
InChI Key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES
[Cl-].[K+]
Conductivity
4950 to 5050 micromhos at 25 deg C
Appearance (Color)
Clear, colorless
Form
liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text