Potassium chloride, 0.159M, Conductivity Standard, Thermo Scientific Chemicals
Potassium chloride, 0.159M, Conductivity Standard, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium chloride, 0.159M, Conductivity Standard, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
500 mL
1 L
Catalog number 042697.K7
also known as 042697-K7
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
Potassium chloride, 0.159M, Conductivity Standard, Thermo Scientific Chemicals
Catalog number042697.K7
Price (USD)/ Each
114.00
-
Add to cart
Specifications
Chemical Name or MaterialPotassium chloride
Name Note20, 000μmho at 25°C
CAS7447-40-7
Quantity4 L
CAS7732-18-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only