Amberlite™ IRA-958(Cl), ion exchange resin, Thermo Scientific Chemicals
Amberlite™ IRA-958(Cl), ion exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlite™ IRA-958(Cl), ion exchange resin, Thermo Scientific Chemicals

Removes Color, organics, and Cyanide
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number 042702.A7
also known as 042702-A7
Price (USD)/ Each
339.00
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
339.00
Add to cart
Amberlite™ IRA-958(Cl), ion exchange resin, Thermo Scientific Chemicals
Catalog number042702.A7
Price (USD)/ Each
339.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAmberlite IRA-958(Cl)
Physical FormBeads
Recommended StorageAmbient temperatures
TSCANo
MDL NumberMFCD00211600
View more
Removal of color, organics, and cyanideAmberlite™ IRA-958(Cl), ion exchange resin is used as an organic scavenger for the adsorption of natural organic matter from surface water. It is an acrylic resin for decolorization and the removal of organic material. Further, it is useful for decolorizing sugar juices and cane syrups, citric acid purification and capturing metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only