Zinc wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Zinc wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 m
Catalog number 042704.G9
also known as 042704-G9
Price (USD)/ Each
276.00
-
Add to cart
Quantity:
25 m
Request bulk or custom format
Price (USD)/ Each
276.00
Add to cart
Zinc wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number042704.G9
Price (USD)/ Each
276.00
-
Add to cart
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.95% (metals basis)
Appearance (Color)White silver
FormWire, 0.5mm (0.02in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only