Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 m
Catalog number 042705.G5
also known as 042705-G5
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
Add to cart
Zinc wire, 1.0mm (0.04in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number042705.G5
Price (USD)/ Each
95.65
Online exclusive
106.00 
Save 10.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialZinc
ColorGray
CAS7440-66-6
AppearanceGray wire
Recommended StorageAmbient temperatures
View more

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only