Polystyrene latex microsphere, 10.0 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals
Polystyrene latex microsphere, 10.0 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polystyrene latex microsphere, 10.0 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 mL
Catalog number 042717.AA
also known as 042717-AA
Price (USD)/ Each
527.65
Online exclusive
586.00 
Save 58.35 (10%)
-
Add to cart
Quantity:
5 mL
Request bulk or custom format
Price (USD)/ Each
527.65
Online exclusive
586.00 
Save 58.35 (10%)
Add to cart
Polystyrene latex microsphere, 10.0 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals
Catalog number042717.AA
Price (USD)/ Each
527.65
Online exclusive
586.00 
Save 58.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA2.5 - 2.8 wt% dispersion in water
FormLiquid dispersion of 10 micron Polystyrene latex microspheres.
Assay from Supplier's CofAParticle Size 9.0 - 11.0 micron
Polystyrene latex microsphere is used as a biocompatible material, especially in dentistry. They are used as insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads. It is also used as extrusion molded foamed packaging, dinnerware, bottles, toys and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only