3-Hydroxy-3-methylbutyric acid, 96%
3-Hydroxy-3-methylbutyric acid, 96%
3-Hydroxy-3-methylbutyric acid, 96%
3-Hydroxy-3-methylbutyric acid, 96%
Thermo Scientific Chemicals

3-Hydroxy-3-methylbutyric acid, 96%

CAS: 625-08-1 | C5H10O3 | 118.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 042722.18
also known as 042722-18
Price (USD)
96.65
Special offer
Online exclusive
Ends: 26-Sep-2025
119.00
Save 22.35 (19%)
Each
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 26-Sep-2025
119.00
Save 22.35 (19%)
Each
Add to cart
Specifications
Beilstein1743952
CAS625-08-1
ChEBICHEBI:37084
Chemical Name or Material3-Hydroxy-3-methylbutyric acid
ColorYellow
View more
Ask our AI about this Product
3-Hydroxy-3-methylbutyric acid is a useful biochemical for synthesis and proteomics research. It is used as an indicator of biotin deficiency. Further, it is used to increase muscle mass and decrease muscle breakdown. In addition to this, it is involved in the protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link