Thermo Scientific Chemicals

Silica gel 60, 0.075-0.2mm (70-200 mesh)

Catalog number: 042723.A1
70/150 Mesh, 1 kg, 500 to 600 m2/g, Each
Thermo Scientific Chemicals

Silica gel 60, 0.075-0.2mm (70-200 mesh)

Catalog number: 042723.A1
70/150 Mesh, 1 kg, 500 to 600 m2/g, Each
Particle Size
Quantity
Surface Area (Metric)
Catalog number: 042723.A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Particle Size
75-200 Microns
Particle Size
60 Angstroms mean pore diameter

Description

Classic column chromatography with no applied pressure (gravity pulled)Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text