Silica gel, wide pore, 150 angstroms
Silica gel, wide pore, 150 angstroms
Thermo Scientific Chemicals

Silica gel, wide pore, 150 angstroms

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
5 kg
Catalog number 042726.30
also known as 042726-30
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
Silica gel, wide pore, 150 angstroms
Catalog number042726.30
Price (USD)/ Each
114.00
-
Add to cart
Chemical Identifiers
CAS63231-67-4
IUPAC Namedihydroxysilanone
Molecular FormulaH2O3Si
InChI KeyIJKVHSBPTUYDLN-UHFFFAOYSA-N
SMILESO[Si](O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Loss on Drying≤2.0%
Particle SizePore diameter (av.): 130-170 angstroms
Particle SizeReport particle size distribution (microns, typical 40-75)
View more
Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only