Thermo Scientific Chemicals

Potassium hydroxide, 0.1N Standardized Solution in methanol, Thermo Scientific Chemicals

Catalog number: 042734.M4
20 L, Each
Thermo Scientific Chemicals

Potassium hydroxide, 0.1N Standardized Solution in methanol, Thermo Scientific Chemicals

Catalog number: 042734.M4
20 L, Each
Quantity
Catalog number: 042734.M4
also known as 042734-M4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-58-3
IUPAC Name
potassium hydroxide
Molecular Formula
HKO
InChI Key
KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES
[OH-].[K+]
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Concentration
0.1N (0.14% uncertainty)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text