Thermo Scientific Chemicals

Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g, Thermo Scientific Chemicals

Catalog number: 042737.30
250 g, Each
Thermo Scientific Chemicals

Silicon(IV) oxide, Amorphous fumed, S.A. 85 to 115m2/g, Thermo Scientific Chemicals

Catalog number: 042737.30
250 g, Each
Quantity
Catalog number: 042737.30
also known as 042737-30
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Surface Area
85-115m²/g

Description

Silicon(IV) oxide, amorphous is used as carriers, processing aids, anti-caking and free-flow agents in animal feed. Defoamer applications such as paint, food, paper, textile and other industrial applications. Synthetic silicon dioxides are used as a rheology control agent in plastics. It is also used to manufacture adhesives, sealants and silicones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 85 to 115m2/g
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text