Thermo Scientific Chemicals

Silicon(IV) oxide, Amorphous fumed, S.A. 175 to 225m2/g, Thermo Scientific Chemicals

Catalog number: 042740.18
50 g, Each
Thermo Scientific Chemicals

Silicon(IV) oxide, Amorphous fumed, S.A. 175 to 225m2/g, Thermo Scientific Chemicals

Catalog number: 042740.18
50 g, Each
Quantity
Catalog number: 042740.18
also known as 042740-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Surface Area
175-2250m²/g

Description

Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 175 to 225m2/g
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text