Thermo Scientific Chemicals

Polystyrene latex microsphere, 1 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals

Catalog number: 042743.4Y
15 mL, Each
Thermo Scientific Chemicals

Polystyrene latex microsphere, 1 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals

Catalog number: 042743.4Y
15 mL, Each
Quantity
Catalog number: 042743.4Y
also known as 042743-4Y
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-53-6
Molecular Formula
(C8H8)n
SMILES
*-CC(-*)C1=CC=CC=C1
MDL Number
MFCD00084450
Form
Liquid dispersion of 1 micron Polystyrene latex microspheres.
Assay from Supplier's CofA
Particle Size 0.90 - 1.10 micron.
Assay from Supplier's CofA
2.5 - 2.8 wt% dispersion in water.

Description

Polystyrene latex microspheres are utilized in flow cytometry, fluorescence microscopy, and as calibration particles. It finds a use in the detection of trace amounts of antigens or antibodies in serum, urine, and cerebrospinal fluids. It is also employed as a stimulus responsive particulate emulsifier for oil-in-water emulsion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text