Cobalt(II) carbonate, 99.5% (metals basis), Thermo Scientific Chemicals
Cobalt(II) carbonate, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt(II) carbonate, 99.5% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 042745.30
also known as 042745-30
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
Add to cart
Cobalt(II) carbonate, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number042745.30
Price (USD)/ Each
250.65
Online exclusive
278.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS513-79-1
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Cobalt(II) carbonate is a precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and is also used in the manufacture of paints and pigments.