Thermo Scientific Chemicals

Alizarin Red S sodium salt, 1% w/v aq. sol.

Catalog number: 042746.AE
100 mL, Each
Thermo Scientific Chemicals

Alizarin Red S sodium salt, 1% w/v aq. sol.

Catalog number: 042746.AE
100 mL, Each
Quantity
Catalog number: 042746.AE
also known as 042746-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
130-22-3
IUPAC Name
sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Molecular Formula
C14H7NaO7S
InChI Key
HFVAFDPGUJEFBQ-UHFFFAOYSA-M
SMILES
[Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O
Concentration
0.8 - 1.2% w/v

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text