Alizarin Red S sodium salt, 1% w/v aq. sol.
Alizarin Red S sodium salt, 1% w/v aq. sol.
Thermo Scientific Chemicals

Alizarin Red S sodium salt, 1% w/v aq. sol.

Transition interval: pH 4.6 (yellow) to pH 6.0 (red)
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number 042746.AP
also known as 042746-AP
Price (USD)/ Each
41.65
Online exclusive
45.80 
Save 4.15 (9%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
45.80 
Save 4.15 (9%)
Add to cart
Alizarin Red S sodium salt, 1% w/v aq. sol.
Catalog number042746.AP
Price (USD)/ Each
41.65
Online exclusive
45.80 
Save 4.15 (9%)
-
Add to cart
Chemical Identifiers
CAS130-22-3
IUPAC Namesodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Molecular FormulaC14H7NaO7S
InChI KeyHFVAFDPGUJEFBQ-UHFFFAOYSA-M
SMILES[Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration0.8 - 1.2% w/v
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only