Thermo Scientific Chemicals
Silica gel, TLC high purity grade, without binder, Thermo Scientific Chemicals
Catalog number: 042756.30
250 g, Each
Thermo Scientific Chemicals
Silica gel, TLC high purity grade, without binder, Thermo Scientific Chemicals
Catalog number: 042756.30
250 g, Each
Quantity
Catalog number: 042756.30
also known as 042756-30
Price (USD)
Quantity
-
Have Questions?
Description
Silica gel, TLC high purity grade, without binder is employed for thin layer chromatography plate preparation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov