Silica gel 60, with fluorescent indicator, 0.060-0.2mm (70-230 mesh), Thermo Scientific Chemicals
Silica gel 60, with fluorescent indicator, 0.060-0.2mm (70-230 mesh), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silica gel 60, with fluorescent indicator, 0.060-0.2mm (70-230 mesh), Thermo Scientific Chemicals

Dry column chromatography without applied pressure (gravity pulled)
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
50 g
250 g
1 kg
Catalog number 042757.A7
also known as 042757-A7
Price (USD)/ Each
1,063.00
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
1,063.00
Add to cart
Silica gel 60, with fluorescent indicator, 0.060-0.2mm (70-230 mesh), Thermo Scientific Chemicals
Catalog number042757.A7
Price (USD)/ Each
1,063.00
-
Add to cart
Specifications
Chemical Name or MaterialSilica gel 60
Physical Form-70+230 Mesh Powder,S.A. 500-600m
Name NoteWith fluorescent indicator
CAS7631-86-9
Health Hazard 1Warning
View more
Silica gel 60 is used as carriers, processing aids, anti-caking and free-flow agents in animal feed. Defoamer applications such as paint, food, paper, textile and other industrial applications. Synthetic silicon dioxides are used as a rheology control agent in plastics. It is also used in dry column chromatography without applied pressure (gravity pulled).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only