Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator, Thermo Scientific Chemicals
Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator, Thermo Scientific Chemicals

TLC plate preparation
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 042764.A1
also known as 042764-A1
Price (USD)/ Each
381.00
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
381.00
Add to cart
Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator, Thermo Scientific Chemicals
Catalog number042764.A1
Price (USD)/ Each
381.00
-
Add to cart
Specifications
Chemical Name or MaterialSilica gel
Physical Form12 Micron APS,S.A. 500-600m
pH6.5 to 7.5
Name NoteWith gypsum binder and fluorescent indicator
CAS63231-67-4
View more
Silica gel is primarily used in plate preparation of thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only