TLC Grade 12 Micron APS Silica Gel
TLC Grade 12 Micron APS Silica Gel
Thermo Scientific Chemicals
TLC Grade 12 Micron APS Silica Gel
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
50 g
Catalog number 042765.30
also known as 042765-30
Price (USD)/ Each
81.30
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
81.30
Add to cart
TLC Grade 12 Micron APS Silica Gel
Catalog number042765.30
Price (USD)/ Each
81.30
-
Add to cart
Specifications
Particle Size12 μm
Pore Size60 Å
GradeTLC
Quantity250 g
Surface Area (Metric)500 to 600 m2/g
View more
TLC plate preparation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only