Silicon(IV) oxide, amorphous fumed, surface treated, S.A. 205-245m2/g, Thermo Scientific Chemicals
Silicon(IV) oxide, amorphous fumed, surface treated, S.A. 205-245m2/g, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon(IV) oxide, amorphous fumed, surface treated, S.A. 205-245m2/g, Thermo Scientific Chemicals

Coatings, thermal insulation, silicones
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 042782.18
also known as 042782-18
Price (USD)/ Each
21.20
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
21.20
Add to cart
Silicon(IV) oxide, amorphous fumed, surface treated, S.A. 205-245m2/g, Thermo Scientific Chemicals
Catalog number042782.18
Price (USD)/ Each
21.20
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area205-245m²/g
Coatings, thermal insulation, silicones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only