Silicon(IV) oxide, Amorphous fumed, Surface treated, S.A. 105 to 130m2/g, Thermo Scientific Chemicals
Silicon(IV) oxide, Amorphous fumed, Surface treated, S.A. 105 to 130m2/g, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon(IV) oxide, Amorphous fumed, Surface treated, S.A. 105 to 130m2/g, Thermo Scientific Chemicals

For Use in Coatings, Adhesives, Sealants
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number 042784.A1
also known as 042784-A1
Price (USD)/ Each
123.00
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
123.00
Add to cart
Silicon(IV) oxide, Amorphous fumed, Surface treated, S.A. 105 to 130m2/g, Thermo Scientific Chemicals
Catalog number042784.A1
Price (USD)/ Each
123.00
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area105-130m²/g
Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 105 to 130m2/g
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only