Thermo Scientific Chemicals

Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: 042804.14
25 g, Each
Thermo Scientific Chemicals

Divinylbenzene, 80%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: 042804.14
25 g, Each
Quantity
Catalog number: 042804.14
also known as 042804-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1321-74-0
IUPAC Name
1,2-diethenylbenzene
Molecular Formula
C10H10
InChI Key
MYRTYDVEIRVNKP-UHFFFAOYSA-N
SMILES
C=CC1=CC=CC=C1C=C
Appearance (Color)
Colorless to yellow
Form
Liquid
Color (APHA)
≤500
Assay (unspecified)
79.0 to 81.0% (mixture of isomers, meta and para)
Stabilizer
900-1100ppm 4-tert-butylcatechol

Description

Divinylbenzene is used as a reactive monomer in polyester resins and for the synthesis of highly crosslinked macroporous styrene as well as a crosslinking agent. It acts as an intermediate in the production of ion exchange resin. Further, it is used in making rubber, petroleum production and polymeric core shell nanoparticles. In addition, it is used to synthesize monodispersed poly divinylbenzene microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text