Thermo Scientific Chemicals

Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: 042805.18
50 g, Each
Thermo Scientific Chemicals

Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: 042805.18
50 g, Each
Quantity
Catalog number: 042805.18
also known as 042805-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1321-74-0
IUPAC Name
1,2-diethenylbenzene
Molecular Formula
C10H10
InChI Key
MYRTYDVEIRVNKP-UHFFFAOYSA-N
SMILES
C=CC1=CC=CC=C1C=C
Form
Liquid
Stabilizer
900 to 1100 ppm TBC
Assay (GC)
≥41.0% Ethyl vinylbenzene
Assay (GC)
55.0-57.0%
Clarity
Clear

Description

Used as a co-monomer in the production of ion exchange beads and gel chromatography. As a monomer in the production of synthetic rubber and casting resins. Also used in polymeric core shell nanoparticles and as a crosslinking agent. It may be used as a monomer for the synthesis of highly crosslinked macroporous styrene/DVB polymers. It may also be used to synthesize monodispersed poly-DVB microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text