Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Crosslinking monomer
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 042805.30
also known as 042805-30
Price (USD)/ Each
59.70
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
59.70
Add to cart
Divinylbenzene, 55%, mixture of isomers, stab. with 1000ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Catalog number042805.30
Price (USD)/ Each
59.70
-
Add to cart
Chemical Identifiers
CAS1321-74-0
IUPAC Name1,2-diethenylbenzene
Molecular FormulaC10H10
InChI KeyMYRTYDVEIRVNKP-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥41.0% Ethyl vinylbenzene
Assay (GC)55.0-57.0%
ClarityClear
FormLiquid
Stabilizer900 to 1100 ppm TBC
Used as a co-monomer in the production of ion exchange beads and gel chromatography. As a monomer in the production of synthetic rubber and casting resins. Also used in polymeric core shell nanoparticles and as a crosslinking agent. It may be used as a monomer for the synthesis of highly crosslinked macroporous styrene/DVB polymers. It may also be used to synthesize monodispersed poly-DVB microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only