Glassy carbon rod, 4mm (0.16in) dia, type 1, Thermo Scientific Chemicals
Glassy carbon rod, 4mm (0.16in) dia, type 1, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon rod, 4mm (0.16in) dia, type 1, Thermo Scientific Chemicals

Used as an electrode material in electrochemistry. It used as high temperature crucibles and as a component of some prosthetic devices. Also used as electrical conductor ion-selective membrane.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mm
100 mm
200 mm
Catalog number 042823.DJ
also known as 042823-DJ
Price (USD)/ Each
270.65
Online exclusive
300.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
50 mm
Request bulk or custom format
Price (USD)/ Each
270.65
Online exclusive
300.00 
Save 29.35 (10%)
Add to cart
Glassy carbon rod, 4mm (0.16in) dia, type 1, Thermo Scientific Chemicals
Catalog number042823.DJ
Price (USD)/ Each
270.65
Online exclusive
300.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 4mm (0.16in) diameter (Glassy Carbon type-1, maximimum service temperature 1000?C).
Assay from Supplier's CofA>99.9%
Appearance (Color)Black
Glassy carbon rod is used as an electrode material in electrochemistry. It used as high temperature crucibles and as a component of some prosthetic devices. Also used as electrical conductor ion-selective membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only