Thermo Scientific Chemicals

Glassy carbon rod, 4mm (0.16in) dia, type 1, Thermo Scientific Chemicals

Catalog number: 042823.DR
200 mm, Each
Thermo Scientific Chemicals

Glassy carbon rod, 4mm (0.16in) dia, type 1, Thermo Scientific Chemicals

Catalog number: 042823.DR
200 mm, Each
Quantity
Catalog number: 042823.DR
also known as 042823-DR
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-44-0
IUPAC Name
carbon
Molecular Formula
C
InChI Key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES
[C]
Appearance (Color)
Black
Form
Rod, 4mm (0.16in) diameter (Glassy Carbon type-1, maximimum service temperature 1000?C).
Assay from Supplier's CofA
>99.9%

Description

Glassy carbon rod is used as an electrode material in electrochemistry. It used as high temperature crucibles and as a component of some prosthetic devices. Also used as electrical conductor ion-selective membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text