Thermo Scientific Chemicals

Antimony(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 042831.04
2 g, Each
Thermo Scientific Chemicals

Antimony(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 042831.04
2 g, Each
Quantity
Catalog number: 042831.04
also known as 042831-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-44-5
IUPAC Name
antimony(3+) triiodide
Molecular Formula
I3Sb
InChI Key
KWQLUUQBTAXYCB-UHFFFAOYSA-K
SMILES
[Sb+3].[I-].[I-].[I-]
Appearance (Color)
Red
Form
Powder
Total Metal Impurities
0.001% max.
Residual water
< 100 ppm

Description

Antimony(III) iodide is used as a dopant in the preparation of thermoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text