Nickel rod, 5mm (0.2in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Nickel rod, 5mm (0.2in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel rod, 5mm (0.2in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
150 mm
50 mm
Catalog number 042835.DO
also known as 042835-DO
Price (USD)/ Each
565.65
Online exclusive
628.00 
Save 62.35 (10%)
-
Add to cart
Quantity:
150 mm
Request bulk or custom format
Price (USD)/ Each
565.65
Online exclusive
628.00 
Save 62.35 (10%)
Add to cart
Nickel rod, 5mm (0.2in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number042835.DO
Price (USD)/ Each
565.65
Online exclusive
628.00 
Save 62.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Commentdiameter 0.48 - 0.52mm
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only