Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells.
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042841.09
also known as 042841-09
Price (USD)/ Each
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7758-95-4
IUPAC Nameλ²-lead(2+) dichloride
Molecular FormulaCl2Pb
InChI KeyHWSZZLVAJGOAAY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities0.001% max.
Appearance (Color)White
Residual water< 100 ppm
Lead chloride is used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. Analytical reagent, preparation of lead salts, as solder and flux. Pattinson's white lead, pigment in white paint, is the basic lead chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only