Magnesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Magnesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042850.06
also known as 042850-06
Price (USD)/ Each
49.65
Online exclusive
55.20 
Save 5.55 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
55.20 
Save 5.55 (10%)
Add to cart
Magnesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number042850.06
Price (USD)/ Each
49.65
Online exclusive
55.20 
Save 5.55 (10%)
-
Add to cart
Chemical Identifiers
CAS7786-30-3
IUPAC Namemagnesium(2+) dichloride
Molecular FormulaCl2Mg
InChI KeyTWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Metal Impurities≤0.01%
FormPowder or crystals or crystalline powder
Magnesium chloride is generally used as a Lewis acid catalyst. It is used as a flocculating agent, analytical reagent, anesthetic for cephalopods, and also in various magnesium cements, and ceramics. It is used for dust control, and road stabilization. It is used in fire extinguishers, polymerase chain reaction (PCR) and hydrogen storage. It is a precursor for magnesium metal and other magnesium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only