Thermo Scientific Chemicals

Cadmium chloride, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog number: 042852.04
2 g, Each
Thermo Scientific Chemicals

Cadmium chloride, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog number: 042852.04
2 g, Each
Quantity
Catalog number: 042852.04
also known as 042852-04
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10108-64-2
IUPAC Name
cadmium(2+) dichloride
Molecular Formula
CdCl2
InChI Key
YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cd++]
Form
Powder
Total Metal Impurities
0.004% max.
Appearance (Color)
White
Residual water
< 100 ppm

Description

Cadmium chloride is used for the preparation of cadmium sulfide, used as Cadmium Yellow, a brilliant-yellow stable inorganic pigment. It can be used for the preparation of organocadmium compounds of the type R2Cd, where R is an aryl or a primary alkyl. It is also used for photocopying, dyeing and electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text