Thermo Scientific Chemicals

Aluminum dihydrogen phosphate, 50% w/w aqueous solution, Thermo Scientific Chemicals

Catalog number: 042858.A1
1 kg, Each
Thermo Scientific Chemicals

Aluminum dihydrogen phosphate, 50% w/w aqueous solution, Thermo Scientific Chemicals

Catalog number: 042858.A1
1 kg, Each
Quantity
Catalog number: 042858.A1
also known as 042858-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13530-50-2
IUPAC Name
aluminium(3+) tridihydrogen phosphate
Molecular Formula
AlH6O12P3
InChI Key
RGPUVZXXZFNFBF-UHFFFAOYSA-K
SMILES
[Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O
Appearance (Color)
Colorless
Form
Liquid
Assay (special)
33.0-37.0% P₂O₅
Assay (special)
7.8-8.8% Al₂O₃
Assay (special)
Report %: Free Acid

Description

Aluminum dihydrogen phosphate is used as intermediates and solid separation agents. It is also used in petroleum, chemical industry, shipbuilding, space technology, electric industry, high-temperature kilns, heat treatment resistance furnaces and electric insulation. Further, it is used as inorganic coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text