Silver wool, 99.9+% (metals basis), Thermo Scientific Chemicals
Silver wool, 99.9+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver wool, 99.9+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 042860.14
also known as 042860-14
Price (USD)/ Each
361.65
Online exclusive
402.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
361.65
Online exclusive
402.00 
Save 40.35 (10%)
Add to cart
Silver wool, 99.9+% (metals basis), Thermo Scientific Chemicals
Catalog number042860.14
Price (USD)/ Each
361.65
Online exclusive
402.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSilver fibres (~0.05mm diameter)
Assay from Suppliers CofA≥99.9% (metals basis)
Appearance (Color)Silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only