Yttrium(III) oxide, REacton™, 99.9999% (REO), Thermo Scientific Chemicals
Yttrium(III) oxide, REacton™, 99.9999% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) oxide, REacton™, 99.9999% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 042864.30
also known as 042864-30
Price (USD)/ Each
819.65
Online exclusive
911.00 
Save 91.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
819.65
Online exclusive
911.00 
Save 91.35 (10%)
Add to cart
Yttrium(III) oxide, REacton™, 99.9999% (REO), Thermo Scientific Chemicals
Catalog number042864.30
Price (USD)/ Each
819.65
Online exclusive
911.00 
Save 91.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-36-9
IUPAC Namediyttrium(3+) trioxidandiide
Molecular FormulaO3Y2
InChI KeyRUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Y+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.0001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only