Thermo Scientific Chemicals

Gadolinium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 042869.18
50 g, Each
Thermo Scientific Chemicals

Gadolinium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 042869.18
50 g, Each
Quantity
Catalog number: 042869.18
also known as 042869-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13450-87-8
IUPAC Name
digadolinium(3+) octahydrate trisulfate
Molecular Formula
Gd2H16O20S3
InChI Key
JBFCVGZGWFIPNK-UHFFFAOYSA-H
SMILES
O.O.O.O.O.O.O.O.[Gd+3].[Gd+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Rare Earth Oxide Impurities
0.1% max.

Description

Gadolinium(III) sulfate is used as analytical reagents, in inorganic synthesis and electronic ceramics. It is also useful in cryogenic research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text