Platinum sponge, 2mm (0.08in) & down, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum sponge, 2mm (0.08in) & down, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum sponge, 2mm (0.08in) & down, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 042871.03
also known as 042871-03
Price (USD)/ Each
280.65
Online exclusive
312.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
280.65
Online exclusive
312.00 
Save 31.35 (10%)
Add to cart
Platinum sponge, 2mm (0.08in) & down, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number042871.03
Price (USD)/ Each
280.65
Online exclusive
312.00 
Save 31.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialPlatinum sponge
CAS7440-06-4
Recommended StorageAmbient temperatures
RTECS NumberTP2160000
EINECS Number231-116-1
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only