Thermo Scientific Chemicals

Ammonium hexachloroosmate(IV), 99.9% (metals basis), Os 42.5% min, Thermo Scientific Chemicals

Catalog number: 042874.06
5 g, Each
Thermo Scientific Chemicals

Ammonium hexachloroosmate(IV), 99.9% (metals basis), Os 42.5% min, Thermo Scientific Chemicals

Catalog number: 042874.06
5 g, Each
Quantity
Catalog number: 042874.06
also known as 042874-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12125-08-5
IUPAC Name
diammonium hexachloroosmiumtetrakis(ylium)
Molecular Formula
Cl6H8N2Os
InChI Key
JRHMYOOMZKAPKT-UHFFFAOYSA-J
SMILES
[NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl
Appearance (Color)
Red to purple
Form
Crystalline
Assay from Supplier's CofA
≥42.5% Os
Purity
≥99.9%

Description

Ammonium hexachloroosmate(IV) acts as a pharmaceutical, organic and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text