Acetonitrile, HPLC Grade (Far UV), 99.8+%, Thermo Scientific Chemicals
Acetonitrile, HPLC Grade (Far UV), 99.8+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetonitrile, HPLC Grade (Far UV), 99.8+%, Thermo Scientific Chemicals

Suitable For Far UV Analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number 042880.K7
also known as 042880-K7
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
Add to cart
Acetonitrile, HPLC Grade (Far UV), 99.8+%, Thermo Scientific Chemicals
Catalog number042880.K7
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSpecification differs for U.S. and non-U.S. material where indicated
Free acid (titration)≤0.001% (10ppm)
Residual water≤0.02% (200ppm)
Optical Absorbance at190nm: ≤1.00AU [U.S. specification] / ≥18% transmittance [non-U.S. specification]
Optical Absorbance at193nm: ≥50% transmittance [non-U.S. specification]
View more
It is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only