Thermo Scientific Chemicals

1,3-Dioxolane, 99.5%, stab., Thermo Scientific Chemicals

Catalog number: 042897.M1
4 x 1 L, Each
Thermo Scientific Chemicals

1,3-Dioxolane, 99.5%, stab., Thermo Scientific Chemicals

Catalog number: 042897.M1
4 x 1 L, Each
Quantity
Catalog number: 042897.M1
also known as 042897-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
646-06-0
IUPAC Name
1,3-dioxolane
Molecular Formula
C3H6O2
InChI Key
WNXJIVFYUVYPPR-UHFFFAOYSA-N
SMILES
C1COCO1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥99.5%
Water Content (Karl Fischer Titration)
≤300 ppm
Peroxide content
≤50 ppm

Description

As a solvent, in Mannich reaction, as an electrolyte in batteries.1,3-Dioxolane is used as a solvent for paints, cellulose esters and in organic synthesis. It is used as an extragent for waxes, fats and oils, stabilizer for halogenated hydrocarbons and co monomer in polyacetals preparation. It is used as finishing agent and swelling agent in textiles. It acts as an intermediate for the preparation of acyclovir and vandetanib. It is an essential ingredient in industrial polymers. In polymerization reactions, it acts as a chain length regulator and a chain transfer agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text