Platinum 13 wt% Rhodium wire, 0.127mm (0.005in) dia, ISA Type R Standard Grade Thermocouple, Thermo Scientific Chemicals
Platinum 13 wt% Rhodium wire, 0.127mm (0.005in) dia, ISA Type R Standard Grade Thermocouple, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum 13 wt% Rhodium wire, 0.127mm (0.005in) dia, ISA Type R Standard Grade Thermocouple, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
25 m
Catalog number 042904.G5
also known as 042904-G5
Price (USD)/ Each
-
Quantity:
5 m
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum 13 wt% Rhodium wire
Name Note0.127mm (0.005 in.) dia., ISA Type R Standard Grade Thermocouple
Recommended StorageAmbient temperatures
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh; 87:13 wt%
TSCAYes