Platinum 6 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Platinum 6 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum 6 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 cm
25 cm
Catalog number 042914.BY
also known as 042914-BY
Price (USD)/ Each
-
Quantity:
100 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum 6 wt% Rhodium wire
Name Note0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple
Recommended StorageAmbient temperatures
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh; 94:6 wt%
TSCAYes
View more
Platinum rhodium wire is used as a catalyst in preparation of nitric acid and in rayon spinnerets. It is used as a device for measuring high temperatures. It serves as a catalyst in the oxidation of the ammonia by atmospheric oxygen to produce nitric oxide, through Haber process.