Silver slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.99% (metals basis), Thermo Scientific Chemicals
Silver slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 Slugs
1 Slug
5 Slugs
Catalog number 042921.QK
also known as 042921-QK
Price (USD)/ Each
476.65
Online exclusive
529.00 
Save 52.35 (10%)
-
Add to cart
Quantity:
25 Slugs
Request bulk or custom format
Price (USD)/ Each
476.65
Online exclusive
529.00 
Save 52.35 (10%)
Add to cart
Silver slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number042921.QK
Price (USD)/ Each
476.65
Online exclusive
529.00 
Save 52.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.99% (metals basis)
Appearance (Color)Silver
FormSlug, 6.35mm (0.25in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only