Copper(II) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals
Copper(II) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number 042925.A1
also known as 042925-A1
Price (USD)/ Each
716.65
Online exclusive
796.00 
Save 79.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
716.65
Online exclusive
796.00 
Save 79.35 (10%)
Add to cart
Copper(II) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number042925.A1
Price (USD)/ Each
716.65
Online exclusive
796.00 
Save 79.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1317-40-4
IUPAC Namecopper(2+) sulfanediide
Molecular FormulaCuS
InChI KeyOMZSGWSJDCOLKM-UHFFFAOYSA-N
SMILES[S--].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Copper(II) sulfide is used in antifouling paints, mixed catalysts and to develop aniline black dyes in textile printing. Further, it is used as a varnish pigment and an oxidizing agent. In addition, it is used as a component of solid state electrodes and thin-film solar cell with cadmium sulfide. It is a moderate conductor of electricity and used in photovoltaics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only