Strontium granules, 20mm max, 99.0% (metals basis), Thermo Scientific Chemicals
Strontium granules, 20mm max, 99.0% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium granules, 20mm max, 99.0% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 042929.06
also known as 042929-06
Price (USD)/ Each
759.65
Online exclusive
844.00 
Save 84.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
759.65
Online exclusive
844.00 
Save 84.35 (10%)
Add to cart
Strontium granules, 20mm max, 99.0% (metals basis), Thermo Scientific Chemicals
Catalog number042929.06
Price (USD)/ Each
759.65
Online exclusive
844.00 
Save 84.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-24-6
IUPAC Namestrontium
Molecular FormulaSr
InChI KeyCIOAGBVUUVVLOB-UHFFFAOYSA-N
SMILES[Sr]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPieces or granules
Appearance (Color)Gold to silver to white
Total Metal Impurities≤1.0%
Appearance (Color)Gold to silver to white
FormPieces or granules
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only