Thermo Scientific Chemicals

Strontium granules, 20mm max, 99.0% (metals basis), Thermo Scientific Chemicals

Catalog number: 042929.14
25 g, Each
Thermo Scientific Chemicals

Strontium granules, 20mm max, 99.0% (metals basis), Thermo Scientific Chemicals

Catalog number: 042929.14
25 g, Each
Quantity
Catalog number: 042929.14
also known as 042929-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-24-6
IUPAC Name
strontium
Molecular Formula
Sr
InChI Key
CIOAGBVUUVVLOB-UHFFFAOYSA-N
SMILES
[Sr]
Form
Pieces or granules
Appearance (Color)
Gold to silver to white
Total Metal Impurities
≤1.0%
Appearance (Color)
Gold to silver to white
Form
Pieces or granules

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text