Lithium isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Lithium isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 042930.04
also known as 042930-04
Price (USD)/ Each
74.50
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
74.50
Add to cart
Lithium isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number042930.04
Price (USD)/ Each
74.50
-
Add to cart
Specifications
Chemical Name or MaterialLithium isopropoxide
CAS2388-10-5
Health Hazard 1H228-H314-H500
Health Hazard 2GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Health Hazard 3P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only