Thermo Scientific Chemicals

Stainless Steel powder, -40+80 mesh, Type 316, Thermo Scientific Chemicals

Catalog number: 042944.A1
1 kg, Each
Thermo Scientific Chemicals

Stainless Steel powder, -40+80 mesh, Type 316, Thermo Scientific Chemicals

Catalog number: 042944.A1
1 kg, Each
Quantity
Catalog number: 042944.A1
also known as 042944-A1
Price (USD)
Quantity
-
Have Questions?
Composition
S: Report
Composition
Si: Report
Form
Powder, -40+80 mesh
Composition
Ni: Report
Composition
C: Report

Description

Stainless Steel powder, Type 316 is a increased resistance to chemical corrosion for the addition of Molybdenum. Also used in a wide variety of home and commercial applications, including, chemical process equipment, food and beverage industry and fasteners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text