Disodium terephthalate, 99+%, Thermo Scientific Chemicals
Disodium terephthalate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Disodium terephthalate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 042946.06
Price (USD)/ Each
25.65
Online exclusive
28.10 
Save 2.45 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
28.10 
Save 2.45 (9%)
Add to cart
Disodium terephthalate, 99+%, Thermo Scientific Chemicals
Catalog number042946.06
Price (USD)/ Each
25.65
Online exclusive
28.10 
Save 2.45 (9%)
-
Add to cart
Specifications
Chemical Name or MaterialDisodium terephthalate
CAS10028-70-3
Recommended StorageAmbient temperatures
EINECS Number233-075-5
TSCANo
View more
Disodium terephthalate and its various derivatives are synthesized via simple acid-base chemistry for anode materials in Na ion batteries. They show excellent electrochemical performance, including little capacity fading over 90 cycles, ideal redox potential, and excellent rate performance, making them promising candidates for Na ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only