N-Methylisobutylamine, 98+%, Thermo Scientific Chemicals
N-Methylisobutylamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methylisobutylamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 042947.06
also known as 042947-06
Price (USD)/ Each
76.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
76.00
Add to cart
N-Methylisobutylamine, 98+%, Thermo Scientific Chemicals
Catalog number042947.06
Price (USD)/ Each
76.00
-
Add to cart
Chemical Identifiers
CAS625-43-4
IUPAC Namemethyl(2-methylpropyl)azanium
Molecular FormulaC5H14N
InChI KeyQKYWADPCTHTJHQ-UHFFFAOYSA-O
SMILESC[NH2+]CC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid
Refractive Index1.3950 to 1.3970
Assay (GC)> 98.0%
N-Methylisobutylamine is used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only